missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium hydroxide, extra pure, 25% solution in water, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 255210025

 View more versions of this product

Catalog No. AC255210025



Specifications

Specifications

Ammonium hydroxide
50ppm max.
NH4OH
ammonia aqueous, ammonia water, ammonia, aqua, ammonia, monohydrate, ammonium hydroxide, ammonium hydroxide nh4 oh, ammonium hydroxide solution, aqua ammonia, aquammonia, household ammonia
[NH4+].[OH-]
50ppm max.
14923
35.05
≥25% (as NH3) (total base)
Glass bottle
0.91
-57.0°C
Colorless
1336-21-6,7732-18-5
H5NO
0.9100g/mL
VHUUQVKOLVNVRT-UHFFFAOYSA-N
azanium;hydroxide
35.046
CHEBI:18219
Liquid
Extra Pure
Solubility in water: soluble.
50ppm max.
38.0°C
2.5L

Chemical Identifiers

1336-21-6
35.046
ammonia aqueous, ammonia water, ammonia, aqua, ammonia, monohydrate, ammonium hydroxide, ammonium hydroxide nh4 oh, ammonium hydroxide solution, aqua ammonia, aquammonia, household ammonia
CHEBI:18219
[NH4+].[OH-]
H5NO
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
azanium;hydroxide
Safety and Handling

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents