Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hydroxide, Environmental Grade, 20-22% NH3

Chemical Identifiers

Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
Purity Grade Environmental Grade
Linear Formula NH4OH
Formula Weight 35.05
MDL Number MFCD00066650
UN Number UN2672
Merck Index 14,494
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38741AP

  SDS
alfa aesar™
38741AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 35.05
Physical Form Liquid, Single Sub-Boiling Quartz Distillation
Odor Ammonia-like
Linear Formula NH4OH
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Concentration 20 to 22% NH3
Density 0.92g/mL
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMMONIA SOLUTION
Chemical Name or Material Ammonium hydroxide
Quantity 500mL
Purity Grade Environmental Grade
RTECS Number BQ9625000
TSCA TSCA
MDL Number MFCD00066650
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN2672
Merck Index 14,494
Color Colorless
CAS 1336-21-6
Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.