Please login to your online account to display your discounted pricing

Ammonium Hydroxide, Certified, 25.0% (v/v) ±1.0%, LabChem™

$76.83 - $76.83

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 7732-18-5
CAS Max % 93.65
CAS 1336-21-6
CAS Max % 6.35
Purity Grade Certified Grade
Linear Formula NH4OH
Formula Weight 35.05
Synonym Ammonia
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC111652

 
lab chem
LC111652
1L Each for $76.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 25.0% ± 1.0% (v/v)
Formula Weight 35.05
Quantity 1L
Viscosity 1.12cSt
Chemical Name or Material Ammonium Hydroxide
Physical Form Liquid
Assay Percent Notes as NH4OH
Density 0.97g/mL
Linear Formula NH4OH
CAS 7732-18-5
Identification Passes Test
Decomposition Information Gaseous Ammonia
Synonym Ammonia
Health Hazard 1 Warning
Packaging Poly Bottle
Odor Ammonia
CAS Max % 93.65
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.
CAS 1336-21-6
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 0.97g/mL
CAS Max % 6.35
Traceability to NIST Traceable to NIST
Color Clear, colorless
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.