Please login to your online account to display your discounted pricing

Ammonium Hydroxide, Certified, 16.1 to 17.3% (v/v), LabChem™

$53.97 - $53.97

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 7732-18-5
CAS Max % 95.8
CAS 1336-21-6
CAS Max % 4.2
Purity Grade Certified Grade
Linear Formula NH4OH
Formula Weight 35.05
Synonym Ammonia
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC111701

 
lab chem
LC111701
Poly Bottle 500mL Each for $53.97
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 35.05
Quantity 500mL
Viscosity 1.08cSt
Health Hazard 1 Warning
Traceability to NIST Traceable to NIST
Chemical Name or Material Ammonium Hydroxide
Decomposition Information Gaseous Ammonia
Physical Form Liquid
Assay 16.1 to 17.3% (v/v)
Assay Percent Notes as NH4OH
Density 0.98g/mL
Linear Formula NH4OH
CAS 7732-18-5
Identification Passes Test
Synonym Ammonia
Packaging Poly Bottle
Odor Ammonia
CAS Max % 95.8
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.
CAS 1336-21-6
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 0.98g/mL
CAS Max % 4.2
Color Clear, colorless
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life.