missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Hydroxide, Certified, 1.00N ±0.02N (1.0M), LabChem™

Supplier:  LabChem LC110804

 View more versions of this product

Catalog No. LC110804



Description

Description

Specifications

Specifications

Ammonium Hydroxide
1336-21-6,7732-18-5
H5NO
MFCD00066650
0.99g/mL
Soluble in water
VHUUQVKOLVNVRT-UHFFFAOYSA-N
amine hydrate
14923
35.05
Passes Test
Poly Bottle
Traceable to NIST
Colorless
98.28,1.72
NH4OH
1.00N ±0.02N (1.0M)
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
4 L
N.O
35.05
CHEBI:18219
Certified
Ammonia
0.99g/mL
Liquid

Chemical Identifiers

1336-21-6
35.05
VHUUQVKOLVNVRT-UHFFFAOYSA-N
14923
amine hydrate
H5NO
MFCD00066650
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
CHEBI:18219
N.O
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions