Please login to your online account to display your discounted pricing

Ammonium hydroxide, for analysis, 20% solution in water, ACROS Organics™


Quantity 2.5L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC390070025

 View more versions of this product

Catalog No. AC390070025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Aluminum (Al) 0.00001 % max.
Chemical Name or Material Ammonium hydroxide
Color Clear green-brown to brown-purple
Quantity 2.5L
CAS 1336-21-6
Antimony (Sb) 0.000002 % max.
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation
Arsenic (As) 0.00001 % max.
CAS 7732-18-5
Health Hazard 3 GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation
IF SWALLOWED: rinse mouth. Do NOT induce vomiting
Wear protective gloves/protective clothing/eye protection/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Avoid breathing dust/fume/gas/mist/vapors/spray

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Appearance Clear liquid
Barium (Ba) 0.00001 % max.
Beryllium (Be) 0.00001 % max.
EINECS Number 215-647-6
Bismuth (Bi) 0.000005 % max.
Cadmium (Cd) 0.00001 % max.
Calcium (Ca) 0.0001 % max.
Carbonate 0.002 % max.
Chloride 0.0001 % max.
Chromium (Cr) 0.000005 % max.
Purity Grade For analysis
Physical Form Solution
Cobalt (Co) 0.00001 % max.
Assay ≥20% (Acidimetry)
Density 0.9200g/mL
Molecular Formula H5NO
Linear Formula NH4OH
Gallium (Ga) 0.000005 % max.
Formula Weight 35.05
MDL Number MFCD00066508
Gold (Au) 0.00001 % max.
Germanium (Ge) 0.000002 % max.
Merck Index 15, 491
Heavy Metals (as Pb) 1ppm max.
Synonym Causes severe skin burns and eye damage
May cause respiratory irritation
Indium (In) 0.000005 % max.
Iron (Fe) 0.00001 % max.
Lead (Pb) 0.000005 % max.
Lithium (Li) 0.000002 % max.
Magnesium (Mg) 0.00005 % max.
Manganese (Mn) 0.000001 % max.
Mercury (Hg) 0.00001 % max.
Molybdenum (Mo) 0.000002 % max.
Nickel (Ni) 0.000005 % max.
Packaging Glass Bottle
Phosphate 0.0001 % max.
Non_volatile Matter 0.002 % max.
Potassium (K) 0.0001 % max.
Platinum (Pt) 0.00001 % max.
Silicon (Si) 0.00002 % max.
Silver (Ag) 0.000002 % max.
Sodium (Na) 0.0002 % max.
Substances Reducing Permanganate 0.001 % max. (as O)
Strontium (Sr) 0.0001 % max.
Sulfide 0.00001 % max.
Thallium (Tl) 0.000002 % max.
Titanium (Ti) 0.000005 % max.
Tin (Sn) 0.000005 % max.
Vanadium (V) 0.000002 % max.
Zirconium (Zr) 0.000005 % max.
Zinc (Zn) 0.00001 % max.
Color Index No. ≤10 APHA

Specifications

Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
CAS 7732-18-5
Purity Grade For analysis
Assay ≥20% (Acidimetry)
Molecular Formula H5NO
Linear Formula NH4OH
Formula Weight 35.05
MDL Number MFCD00066508
Accessories

Danger

GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation
IF SWALLOWED: rinse mouth. Do NOT induce vomiting
Wear protective gloves/protective clothing/eye protection/face protection
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
Avoid breathing dust/fume/gas/mist/vapors/spray

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage
May cause respiratory irritation

SDS & Safety Documents