Please login to your online account to display your discounted pricing

Ammonium Hydroxide, ACS Reagent Grade, 28.0-30.0% as NH3, Ricca Chemical


Quantity 2.50L
Packaging Glass

Manufacturer: ricca chemical company  RIRABA00202.5D1

 View more versions of this product

Catalog No. RABA002025D

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Liquid
Color Clear colorless
Quantity 2.50L
Concentration or Composition Notes 28.0-30.0% as NH3
CAS 1336-21-6
Chemical Name or Material Ammonium Hydroxide
CAS Min % 98.0
Packaging Glass
CAS Max % 100.0
CAS 7732-18-5 (Water, Deionized)

Specifications

Chemical Name or Material Ammonium Hydroxide
CAS 1336-21-6
CAS Min % 98.0
CAS Max % 100.0
CAS 7732-18-5 (Water, Deionized)
Purity Grade ACS Reagent Grade
Physical Form Liquid