Please login to your online account to display your discounted pricing

Ammonium hydroxide, ACS reagent, 28-30% solution in water, ACROS Organics™


Quantity 500mL
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  423305000

 View more versions of this product

Catalog No. AC423305000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Free From Suspended Matter or Sediment
Formula Weight 35.05
Quantity 500mL
Color Colorless
Fieser 05,15
Merck Index 15, 491
EINECS Number 215-647-6
TSCA TSCA
Assay 28.0 to 30.0% (HCl-titration (as NH3))
Density 0.9100g/mL
RTECS Number BQ9625000
Carbon Dioxide 0.002% max.
Chlorine (Cl) 0.5ppm max.
Additional Information Vapor Pressure: 500hPa at 20°C
MDL Number MFCD00066650
Heavy Metals (as Pb) 0.5ppm max.
Iron (Fe) 0.2ppm max.
Health Hazard 1 Danger
Packaging Glass Bottle
Phosphate 2ppm max.
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.
Nitrate 2ppm max.
CAS 7732-18-5
Health Hazard 3 GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Residue after Ignition 0.002% max.
Substances Reducing Permanganate Passes Test
Sulfate 2ppm max.
Chemical Name or Material Ammonium hydroxide
Molecular Formula H5NO
Linear Formula NH4OH
CAS 1336-21-6

Specifications

Chemical Name or Material Ammonium hydroxide
CAS 1336-21-6
CAS 7732-18-5
Purity Grade ACS Reagent Grade
Assay 28.0 to 30.0% (HCl-titration (as NH3))
Molecular Formula H5NO
Linear Formula NH4OH
Formula Weight 35.05
MDL Number MFCD00066650
Accessories

Danger

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May cause respiratory irritation.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.

SDS & Safety Documents