Please login to your online account to display your discounted pricing

Ammonium Hydroxide ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 1336-21-6
Purity Grade ACS Grade
Molecular Formula NH4OH
Formula Weight 35.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK6665212X6

 
avantor performance
666546
SAFEMOR 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6665500

 
avantor performance
666514
Glass 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ammonium Hydroxide
Quantity 2.5L
CAS 1336-21-6
Purity Grade ACS Grade
Molecular Formula NH4OH
Formula Weight 35.05
Packaging SAFEMOR