Please login to your online account to display your discounted pricing

Ammonium Hydroxide, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$141.87 - $589.30

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 1336-21-6
Name Note 5N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NH4OH
Formula Weight 35.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002297

 
avantor performance
560402
Narrow Mouth Polyethylene Bottle 1L Each for $141.87

02002299

 
avantor performance
560403
Cubetainer 4L Each for $589.30
Description & Specifications

Specifications

Formula Weight 35.05
Quantity 1L
Name Note 5N Volumetric Solution
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide
CAS 1336-21-6
Molecular Formula NH4OH
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle