missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ammonium Hydroxide, 25% (v/v), Ricca Chemical
$186.25 - $254.69
Chemical Identifiers
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide |
Specifications
Alkaline | |
7732-18-5 | |
dihydrogen oxide, dihydrogen monoxide | |
1 L | |
Ammonia | |
Liquid |
Colorless | |
25% (v/v) Aqueous Solution (1 + 3) | |
Solubility in Water: Infinite | |
25% | |
Natural Poly Bottle |
Safety and Handling
Recommended Storage : Corrosive