Please login to your online account to display your discounted pricing

Ammonium Hydroxide, 2.00 Normal, Ricca Chemical

$53.12 - $53.12

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 7732-18-5
CAS Min % 87.0
CAS Max % 91.0
CAS 1336-21-6
CAS Min % 10.83
CAS Max % 11.27
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

643-16

  SDS
ricca chemical company
RI64316
Poly natural 500mL Each for $53.12

643-32

  SDS
ricca chemical company
RI64332
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Concentration or Composition Notes 2.00Normal
Chemical Name or Material Ammonium Hydroxide
Packaging Poly natural
CAS 1336-21-6
CAS Min % 87.0
CAS Max % 91.0
CAS Min % 10.83
CAS Max % 11.27
CAS 7732-18-5