Please login to your online account to display your discounted pricing

Ammonium Hydroxide, 0.2, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$139.46 - $2,153.69

Chemical Identifiers

Chemical Name or Material Ammonium Hydroxide
CAS 1336-21-6
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula NH4OH
Formula Weight 35.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002300

 
avantor performance
938003
Pressure Bottom Poly Bottle 4L Each for $2,153.69

02002301

 
avantor performance
938001
Poly Bottle 500mL Each for $139.46
Description & Specifications

Specifications

Formula Weight 35.05
Quantity 4L
Chemical Name or Material Ammonium Hydroxide
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula NH4OH
CAS 1336-21-6
Assay 20%
Packaging Pressure Bottom Poly Bottle
Specific Gravity 1L = 0.93kg

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use