Learn More
Ammonium hydrogen sulfate, 99%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389412500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
350.0°C | |
147.0°C | |
White | |
250 g | |
H5NO4S | |
MFCD00051073 | |
ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4 | |
BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
azanium;hydrogen sulfate | |
24655 | |
99% | |
Glass bottle |
Ammonium hydrogen sulfate | |
7803-63-6 | |
Crystalline Powder | |
99% | |
NH4HSO4 | |
15, 497 | |
Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol, acetone and, pyridine | |
[NH4+].OS([O-])(=O)=O | |
115.10 | |
115.11 | |
Analytical | |
0.01% max. (600°C) |
Chemical Identifiers
7803-63-6 | |
115.10 | |
BIGPRXCJEDHCLP-UHFFFAOYSA-N | |
24655 |
H5NO4S | |
MFCD00051073 | |
ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4 | |
[NH4+].OS([O-])(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 232-265-5
RUO – Research Use Only