Learn More
Ammonium hydrogen phosphate, tech., P{2}O{5} 53% min, Thermo Scientific Chemicals
Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc
$119.08 - $316.93
Chemical Identifiers
CAS | 7783-28-0 |
---|---|
Molecular Formula | H9N2O4P |
Molecular Weight (g/mol) | 132.06 |
MDL Number | MFCD00010891 |
InChI Key | MNNHAPBLZZVQHP-UHFFFAOYSA-N |
Synonym | diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate |
PubChem CID | 24540 |
ChEBI | CHEBI:63051 |
SMILES | N.N.OP(O)(O)=O |
Description
Ammonium hydrogen phosphate is used as a fertilizer, as a fire retardant, as a yeast nutrient in winemaking and brewing mead, as an additive in some brands of cigarettes, in purifying sugar and to control precipitation of alkali-soluble and acid-insoluble colloidal dyes on wool.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-28-0 | |
132.06 | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
24540 | |
N.N.OP(O)(O)=O |
H9N2O4P | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
CHEBI:63051 |
Specifications
155°C (decomposition) | |
White | |
1 kg | |
MFCD00010891 | |
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate | |
MNNHAPBLZZVQHP-UHFFFAOYSA-N | |
diazanium;hydrogen phosphate | |
24540 | |
132.06 | |
P2O5 53% min. | |
Ammonium hydrogen phosphate |
7783-28-0 | |
Powder | |
H9N2O4P | |
14,542 | |
Freely soluble in water; Insoluble in alcohol, acetone | |
N.N.OP(O)(O)=O | |
132.06 | |
CHEBI:63051 | |
Technical | |
1.619 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-987-8
RTECSNumber : TB9385000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only