missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hydrogen phosphate, tech., P2O5 53% min

Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc

Manufacturer:  Alfa Aesar™ 10301A7

 View more versions of this product

Catalog No. AA10301A7



Specifications

Specifications

Ammonium hydrogen phosphate
H9N2O4P
14,542
Freely soluble in water; Insoluble in alcohol, acetone
[NH4+].[NH4+].OP(=O)([O-])[O-]
132.056
CHEBI:63051
Powder
P2O5 53% min.
−100 Mesh
155°C (decomposition)
7783-28-0
MFCD00010891
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
MNNHAPBLZZVQHP-UHFFFAOYSA-N
diazanium;hydrogen phosphate
24540
132.06
Technical
1.619g/mL
White
5kg

Chemical Identifiers

7783-28-0
132.056
MNNHAPBLZZVQHP-UHFFFAOYSA-N
24540
diazanium;hydrogen phosphate
H9N2O4P
MFCD00010891
diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
CHEBI:63051
[NH4+].[NH4+].OP(=O)([O-])[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TB9385000

TSCA : TSCA

Documents