Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hydrogen phosphate, 98%

$52.88 - $52.88

Chemical Identifiers

Chemical Name or Material Ammonium hydrogen phosphate
CAS 7783-28-0
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
Formula Weight 132.06
MDL Number MFCD00010891
Merck Index 14,542
Synonym Diammonium hydrogen phosphate; Phosphoric acid diammonium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1741636

  SDS
alfa aesar™
A1741636
500g Each for $52.88
Description & Specifications

Specifications

Formula Weight 132.06
Chemical Name or Material Ammonium hydrogen phosphate
TSCA TSCA
Assay 98%
Molecular Formula H9N2O4P
Linear Formula (NH4)2HPO4
MDL Number MFCD00010891
CAS 7783-28-0
Merck Index 14,542
Synonym Diammonium hydrogen phosphate; Phosphoric acid diammonium salt
Melting Point 155°C (dec.)
Quantity 500g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TB9385000

Applications
It is commonly used in fertilizer as plant food as well as a yeast nutrient. Ammonium phosphate dibasic has been used to prepare multi-shell nanoparticles to increase transfection efficiency.

Solubility
Soluble in water

Notes
Hygroscopic. Store away from oxidizing agents, acids.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.