missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium hydrogen carbonate, 99.0%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 014249A1

 View more versions of this product

Catalog No. AA14249A1


Description

Description

In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
  • Also used for compost treatment to accelerate decomposition
Specifications

Specifications

Ammonium hydrogen carbonate
1066-33-7
Powder
C3H8N2O9Zn2
MFCD00012138
14,497
Soluble in cold water but decomposed by hot water; Insoluble in alcohol and acetone.
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
346.86
79.06
Like ammonia
∼60°C (decomposition)
White
1 kg
(NH4)HCO3
4329606
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
QOPSSYZLVIENBR-UHFFFAOYSA-J
azanium;hydrogen carbonate
14013
99.0%
1.586 g/mL

Chemical Identifiers

1066-33-7
346.86
QOPSSYZLVIENBR-UHFFFAOYSA-J
14013
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
C3H8N2O9Zn2
MFCD00012138
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
azanium;hydrogen carbonate
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-911-5

RTECSNumber : BO8600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only