Learn More
Ammonium hydrogen carbonate, 99.0%, Thermo Scientific Chemicals
$82.23 - $202.92
Chemical Identifiers
CAS | 1066-33-7 |
---|---|
Molecular Formula | C3H8N2O9Zn2 |
Molecular Weight (g/mol) | 346.86 |
MDL Number | MFCD00012138 |
InChI Key | QOPSSYZLVIENBR-UHFFFAOYSA-J |
Synonym | ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 |
PubChem CID | 14013 |
IUPAC Name | azanium;hydrogen carbonate |
SMILES | [NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
Description
In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
- Also used for compost treatment to accelerate decomposition
Chemical Identifiers
1066-33-7 | |
346.86 | |
QOPSSYZLVIENBR-UHFFFAOYSA-J | |
14013 | |
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
C3H8N2O9Zn2 | |
MFCD00012138 | |
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
azanium;hydrogen carbonate |
Specifications
∼60°C (decomposition) | |
White | |
1 kg | |
(NH4)HCO3 | |
4329606 | |
ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 | |
QOPSSYZLVIENBR-UHFFFAOYSA-J | |
azanium;hydrogen carbonate | |
14013 | |
99.0% | |
1.586 g/mL |
1066-33-7 | |
Powder | |
C3H8N2O9Zn2 | |
MFCD00012138 | |
14,497 | |
Soluble in cold water but decomposed by hot water; Insoluble in alcohol and acetone. | |
[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
346.86 | |
79.06 | |
Like ammonia | |
Ammonium hydrogen carbonate |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 213-911-5
RTECSNumber : BO8600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only