Please login to your online account to display your discounted pricing

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis), ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  194350100

 View more versions of this product

Catalog No. AC194350100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder, Crystals, and/or Chunks
Formula Weight 197.97
Quantity 10g
Color White
Purity Grade For trace metal basis analysis
Chemical Name or Material Ammonium hexafluorotitanate(IV)
Packaging Glass bottle
Assay Percent Range 99.99%
Molecular Formula H8F6N2Ti
Linear Formula (NH4)2TiF6
MDL Number MFCD00010888
CAS 16962-40-6
Total Trace Metal Impurities 0.01% max.
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ammonium hexafluorotitanate(IV)
CAS 16962-40-6
Purity Grade For trace metal basis analysis
Assay Percent Range 99.99%
Molecular Formula H8F6N2Ti
Linear Formula (NH4)2TiF6
Formula Weight 197.97
MDL Number MFCD00010888
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents