Learn More
Ammonium hexafluorosilicate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 194002500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Ammonium hexafluorosilicate | |
White | |
250 g | |
H8F6N2Si | |
15,524 | |
Solubility in water: 18.6 g/100mL (17°C). | |
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
178.15 | |
178.15 | |
Trace Metal Basis | |
2.01 |
16919-19-0 | |
Crystalline Powder and/or Chunks | |
99.999% | |
(NH4)2SiF6 | |
ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate | |
ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
diazanium;hexafluorosilicon(2-) | |
28145 | |
99.999% | |
Glass bottle | |
2.0100g/mL |
Chemical Identifiers
16919-19-0 | |
178.15 | |
ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate | |
diazanium;hexafluorosilicon(2-) |
H8F6N2Si | |
ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
28145 | |
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protectio
GHS Signal Word: Danger
EINECSNumber : 240-968-3
RUO – Research Use Only