missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexafluorosilicate, 98%

Etching glass, in soldering flux

Manufacturer:  Alfa Aesar™ 69100A1

 View more versions of this product

Catalog No. AA69100A1



Specifications

Specifications

Ammonium hexafluorosilicate
F6H8N2Si
UN2854
ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
28145
Powder
1kg
16919-19-0
MFCD00010887
14,527
Slightly soluble in water; Insoluble in alcohol, acetone
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
178.153
178.14
98%

Chemical Identifiers

16919-19-0
178.153
ITHIMUMYFVCXSL-UHFFFAOYSA-P
28145
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
F6H8N2Si
MFCD00010887
ammonium hexafluorosilicate, ammonium fluosilicate, ammonium fluorosilicate, ammonium silicofluoride, cryptohalite, caswell no. 043, diammonium fluosilicate, diammonium fluorosilicate, ammonium silicon fluoride, diammonium hexafluorosilicate
diazanium;hexafluorosilicon(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUOROSILICATE

TSCA : TSCA

SDS Documents