Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexafluoroaluminate, 99.8% (metals basis)

Preparation of aluminum fluoride

Chemical Identifiers

Chemical Name or Material Ammonium hexafluoroaluminate
CAS 7784-19-2
Linear Formula (NH4)3AlF6
Formula Weight 195.09g/mol
MDL Number MFCD00042144
UN Number UN3288
Merck Index 14,524
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3912806

 
alfa aesar™
39128-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3912814

 
alfa aesar™
39128-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 1.78g/mL
Mesh Size −80 Mesh
Odor Odorless
Color White
Quantity 5g
Chemical Name or Material Ammonium hexafluoroaluminate
CAS 7784-19-2
Physical Form Powder
Assay 99.8%
Assay Percent Notes (metals basis)
Linear Formula (NH4)3AlF6
Formula Weight 195.09g/mol
MDL Number MFCD00042144
UN Number UN3288
Merck Index 14,524
Solubility Information Freely soluble in water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
RTECS Number BD0050000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.