Please login to your online account to display your discounted pricing

Ammonium hexafluoroaluminate 95%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC383320500

 View more versions of this product

Catalog No. AC383320500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 195.09
Quantity 50g
Color White
Merck Index 15, 521
Chemical Name or Material Ammonium hexafluoroaluminate
Identification Confirmed (ICP/AAS)
Packaging Glass bottle
Molecular Formula H12AlF6N3
Assay Percent Range 95%
MDL Number MFCD00042144
CAS 7784-19-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Fatal if inhaled.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 94% min. (total fluoride)

Specifications

Chemical Name or Material Ammonium hexafluoroaluminate
Assay Percent Range 95%
Molecular Formula H12AlF6N3
Formula Weight 195.09
MDL Number MFCD00042144
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Fatal if inhaled.
Toxic if swallowed.

SDS & Safety Documents