Please login to your online account to display your discounted pricing

Ammonium hexafluoroaluminate 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ammonium hexafluoroaluminate
Assay Percent Range 95%
Molecular Formula H12AlF6N3
Formula Weight 195.09
MDL Number MFCD00042144
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC383322500

  SDS
thermo fisher scientific
AC383322500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC383320500

  SDS
thermo fisher scientific
AC383320500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 195.09
Quantity 250g
Color White
Merck Index 15, 521
Identification Confirmed (ICP/AAS)
Chemical Name or Material Ammonium hexafluoroaluminate
Molecular Formula H12AlF6N3
MDL Number MFCD00042144
Packaging Glass bottle
Elemental Analysis 94% min. (total fluoride)
Assay Percent Range 95%
CAS 7784-19-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Fatal if inhaled.
Toxic if swallowed.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Fatal if inhaled.
Toxic if swallowed.