missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell Fluka™ Ammonium Hexacyanoferrate(Ii) Hydrate, ≥92.0% anhydrous basis (RT), Honeywell™ Fluka™

Spot test reagent, for the determination of Ca,≥92.0% anhydrous basis (RT), ≤15% water

$209.93 - $1,205.63

Chemical Identifiers

CAS 14481-29-9
Molecular Formula C6H16FeN10
Molecular Weight (g/mol) 284.109
MDL Number MFCD00149957
InChI Key ZXQVPEBHZMCRMC-UHFFFAOYSA-R
Synonym ammonium ferrocyanide, ammonium hexacyanoferrate ii, ammonium hexacyanoferrate ii, ammoniumferrocyanid, tetraammonium ferrocyanide, ammonium hexacyanoferrate 4-, tetraammonium hexacyanidoferrate, ammonium hexacyanidoferrate ii, ammonium hexacyanidoferrate 4-, nh4 4 fe cn 6
PubChem CID 17756740
IUPAC Name tetraazanium;iron(2+);hexacyanide
SMILES [C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[NH4+].[NH4+].[NH4+].[Fe+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044618 View Documents Honeywell Fluka™
09810250G
250g Poly Bottle Each for $1,205.63
 
60044619 View Documents Honeywell Fluka™
0981050G
50g Poly Bottle Each for $209.93
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

14481-29-9
284.109
ZXQVPEBHZMCRMC-UHFFFAOYSA-R
17756740
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[NH4+].[NH4+].[NH4+].[Fe+2]
C6H16FeN10
MFCD00149957
ammonium ferrocyanide, ammonium hexacyanoferrate ii, ammonium hexacyanoferrate ii, ammoniumferrocyanid, tetraammonium ferrocyanide, ammonium hexacyanoferrate 4-, tetraammonium hexacyanidoferrate, ammonium hexacyanidoferrate ii, ammonium hexacyanidoferrate 4-, nh4 4 fe cn 6
tetraazanium;iron(2+);hexacyanide

Specifications

14481-29-9
C6H16FeN10
MFCD00149957
Powder
ZXQVPEBHZMCRMC-UHFFFAOYSA-R
tetraazanium;iron(2+);hexacyanide
17756740
≥92.0% anhydrous basis (RT)
Green to White
Ammonium Hexacyanoferrate(II) Hydrate
Spot test reagent, for determination of Ca
(NH4)4[Fe(CN)6] · xH2O
NONH for all modes of transport
ammonium ferrocyanide, ammonium hexacyanoferrate ii, ammonium hexacyanoferrate ii, ammoniumferrocyanid, tetraammonium ferrocyanide, ammonium hexacyanoferrate 4-, tetraammonium hexacyanidoferrate, ammonium hexacyanidoferrate ii, ammonium hexacyanidoferrate 4-, nh4 4 fe cn 6
[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[NH4+].[NH4+].[NH4+].[NH4+].[Fe+2]
284.109
284.10g/mol (anhydrous basis)
Poly Bottle
250g
Safety and Handling

Safety and Handling

P280

H302 + H312 + H332

EINECSNumber : 238-476-9