Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachlorostannate(IV), 98%

$37.36 - $470.11

Chemical Identifiers

Chemical Name or Material Ammonium hexachlorostannate(IV)
CAS 16960-53-5
Molecular Formula (NH4)2SnCl6
Formula Weight 367.48
MDL Number MFCD00036340
Synonym Stannic ammonium chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA151520B

  SDS
alfa aesar™
A151520B
Each for $470.11

AAA1515218

  SDS
alfa aesar™
A1515218
Each for $37.36
Description & Specifications

Specifications

Formula Weight 367.48
Chemical Name or Material Ammonium hexachlorostannate(IV)
TSCA TSCA
Assay 98%
Molecular Formula (NH4)2SnCl6
MDL Number MFCD00036340
CAS 16960-53-5
Synonym Stannic ammonium chloride
Quantity 1000g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.