Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
CAS 15336-18-2
Purity Grade Premion™
Formula Weight 369.74 (Anhydrous)
MDL Number MFCD00003394
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1181201

  SDS
alfa aesar™
1181201
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1181203

  SDS
alfa aesar™
1181203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1181206

  SDS
alfa aesar™
1181206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 369.74 (Anhydrous)
Chemical Name or Material Ammonium hexachlororhodate(III) hydrate
TSCA TSCA
Purity Grade Premion™
Physical Form Powder
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00003394
CAS 15336-18-2
Color Dark red
Quantity 0.25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.