missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroplatinate(IV), Pt 43.4% min

Electroplating of platinum, manufacturing of platinum sponge

Manufacturer:  Alfa Aesar™ 1288203

 View more versions of this product

Catalog No. AA1288203



Specifications

Specifications

Ammonium hexachloroplatinate(IV)
Cl6H8N2Pt
UN3288
ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
PCCGQTHFYHJATL-UHFFFAOYSA-J
diazanium;hexachloroplatinum(2-)
16211460
443.89
Pt 43.4% min.
Hygroscopic
1g
16919-58-7
MFCD00010886
14,548
Slightly soluble in water; Insoluble in alcohol
[NH4+].[NH4+].Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl
443.862
CHEBI:59604
Powder
3.06g/mL
Yellow

Chemical Identifiers

16919-58-7
443.862
PCCGQTHFYHJATL-UHFFFAOYSA-J
16211460
diazanium;hexachloroplatinum(2-)
Cl6H8N2Pt
MFCD00010886
ammonium hexachloroplatinate iv, ammonium chloroplatinate, ammonium hexachloroplatinate, ammonium platinic chloride, ammonium platinum chloride, diammonium hexachloroplatinate 2-, diammonium platinum hexachloride, ammoniumhexachloroplatinate, epitope id:151516, platinum iv-ammonium chloride
CHEBI:59604
[NH4+].[NH4+].Cl[Pt-2](Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H372-H318-H290-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

RTECSNumber : BP5425000

TSCA : TSCA

Documents