Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloropalladate(IV), 99.9% (metals basis), Pd 29%

Chemical Identifiers

Chemical Name or Material Ammonium hexachloropalladate(IV)
CAS 19168-23-1
Formula Weight 355.2
MDL Number MFCD00015959
UN Number UN3077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1104204

  SDS
alfa aesar™
1104204
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1104209

  SDS
alfa aesar™
1104209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 355.2
Chemical Name or Material Ammonium hexachloropalladate(IV)
TSCA TSCA
Odor Odorless
Physical Form Crystalline
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00015959
CAS 19168-23-1
UN Number UN3077
Color Red-orange
Quantity 2g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Pd 29%
Density 2.42g/mL
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number BP5390000
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.