missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloropalladate(IV), 99.9% (metals basis), Pd 29%

Manufacturer:  Alfa Aesar™ 1104209

 View more versions of this product

Catalog No. AA1104209



Specifications

Specifications

Ammonium hexachloropalladate(IV)
(metals basis)
MFCD00015959
ammonium chloropalladate nh4 2pdcl6
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl
355.198
CHEBI:60124
Crystalline
Pd 29%
Odorless
Red to Orange
19168-23-1
Cl6H8N2Pd
UN3077
LOLIPEAFAJNGJM-UHFFFAOYSA-J
diazanium;hexachloropalladium(2-)
62732
355.2
99.9%
2.42g/mL
Hygroscopic
10g

Chemical Identifiers

19168-23-1
355.198
LOLIPEAFAJNGJM-UHFFFAOYSA-J
62732
diazanium;hexachloropalladium(2-)
Cl6H8N2Pd
MFCD00015959
ammonium chloropalladate nh4 2pdcl6
CHEBI:60124
[NH4+].[NH4+].Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H317-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P310-P362-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : BP5390000

TSCA : TSCA

Documents