Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min

Chemical Identifiers

Chemical Name or Material Ammonium hexachloroiridate(IV)
CAS 16940-92-4
Purity Grade Premion™
Formula Weight 441.02
MDL Number MFCD00010881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1071277

  SDS
alfa aesar™
1071277
0.1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1071203

  SDS
alfa aesar™
1071203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1071206

  SDS
alfa aesar™
1071206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 441.02
Physical Form Crystalline Powder
Color Red-black
Health Hazard 2 GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.
TSCA TSCA
Quantity 0.1g
MDL Number MFCD00010881
Assay Percent Notes (metals basis)
Concentration Ir 41% min.
Density 2.86g/mL
Health Hazard 1 Warning
Chemical Name or Material Ammonium hexachloroiridate(IV)
CAS 16940-92-4
Health Hazard 3 GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
RTECS Number BP5295000
Purity Grade Premion™
Assay 99.994%
Safety and Handling

Warning

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.