missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroiridate(IV), Premion™, 99.994% (metals basis), Ir 41% min

Manufacturer:  Alfa Aesar™ 1071277

 View more versions of this product

Catalog No. AA1071277



Specifications

Specifications

Ammonium hexachloroiridate(IV)
(metals basis)
MFCD00010881
LWNOUTCTZQNGEN-UHFFFAOYSA-J
diazanium;hexachloroiridium(2-)
16211476
441.02
99.994%
Ir 41% min.
Hygroscopic
0.1g
16940-92-4
Cl6H8IrN2
ammonium chloroiridate
[NH4+].[NH4+].Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl
440.995
CHEBI:60132
Crystalline Powder
Premion™
2.86g/mL
Black-Red

Chemical Identifiers

16940-92-4
440.995
LWNOUTCTZQNGEN-UHFFFAOYSA-J
16211476
diazanium;hexachloroiridium(2-)
Cl6H8IrN2
MFCD00010881
ammonium chloroiridate
CHEBI:60132
[NH4+].[NH4+].Cl[Ir-2](Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BP5295000

TSCA : TSCA

Documents