Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroiridate(IV), 99% (metals basis), Ir 43% min

Chemical Identifiers

Chemical Name or Material Ammonium hexachloroiridate(IV)
CAS 16940-92-4
Formula Weight 441.02
MDL Number MFCD00010881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1232603

  SDS
alfa aesar™
1232603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1232606

  SDS
alfa aesar™
1232606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 441.02
MDL Number MFCD00010881
Chemical Name or Material Ammonium hexachloroiridate(IV)
Sensitivity Hygroscopic
Concentration Ir 43% min.
RTECS Number BP5295000
CAS 16940-92-4
Assay 99%
Assay Percent Notes (metals basis)
Health Hazard 3 GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Color Red-black
Density 2.86g/mL
TSCA TSCA
Quantity 1g
Health Hazard 2 GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.
Safety and Handling

Warning

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.