Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroiridate(III) hydrate, Premion™, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
CAS 15752-05-3
Purity Grade Premion™
Formula Weight 459.06 (Anhydrous)
MDL Number MFCD00003393
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1071303

 
alfa aesar™
1071303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1071306

 
alfa aesar™
1071306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 459.06 (Anhydrous)
Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
TSCA TSCA
Odor Odorless
Purity Grade Premion™
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
MDL Number MFCD00003393
CAS 15752-05-3
Color Green
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.