Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexachloroiridate(III) hydrate

Chemical Identifiers

Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
Formula Weight 459.06 (Anhydrous)
MDL Number MFCD00003393
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1263203

  SDS
alfa aesar™
1263203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1263206

  SDS
alfa aesar™
1263206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 459.06 (Anhydrous)
Chemical Name or Material Ammonium hexachloroiridate(III) hydrate
TSCA TSCA
Odor Odorless
Sensitivity Hygroscopic
MDL Number MFCD00003393
Color Green
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.