Alfa Aesar™ Ammonium hexachloroiridate(III) hydrate
Manufacturer:  Alfa Aesar™ 1263203
 View more versions of this product
Catalog No. AA1263203
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Ammonium hexachloroiridate(III) hydrate
Cl6H14IrN3O
ammonium hexachloroiridate iii hydrate, ammonium hexachloroiridate iii hydrate, triammonium hexachloroiridiumtriuide hydrate
[NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
477.049
459.06 (Anhydrous)
Odorless
Green
29796-57-4
MFCD00003393
LJHBEBFYNMDGCZ-UHFFFAOYSA-K
triazanium;hexachloroiridium(3-);hydrate
18366487
Crystalline Powder
Hygroscopic
1g
Chemical Identifiers
29796-57-4
477.049
LJHBEBFYNMDGCZ-UHFFFAOYSA-K
18366487
[NH4+].[NH4+].[NH4+].O.Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
Cl6H14IrN3O
MFCD00003393
ammonium hexachloroiridate iii hydrate, ammonium hexachloroiridate iii hydrate, triammonium hexachloroiridiumtriuide hydrate
triazanium;hexachloroiridium(3-);hydrate
Safety and Handling
Safety and Handling
GHS H Statement
H290-H302
May be corrosive to metals.
Harmful if swallowed.
GHS P Statement
P234-P264-P270-P301+P312-P406-P501a
Keep only in original container.
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA
Documents