Please login to your online account to display your discounted pricing

Ammonium hexabromoplatinate(IV), 99.99%, (trace metal basis), ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ammonium hexabromoplatinate(IV)
Name Note 99.99%
CAS 17363-02-9
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula H8Br6N2Pt
Linear Formula (NH4)2PtBr6
Formula Weight 710.61
MDL Number MFCD00044261
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC193740010

  SDS
thermo fisher scientific
AC193740010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 710.61
Physical Form Crystalline Powder
Purity Grade Trace Metal Basis
Quantity 1g
Color Red
TSCA TSCA
EINECS Number 241-394-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Ammonium hexabromoplatinate(IV)
Assay Percent Range 99.99%
Molecular Formula H8Br6N2Pt
MDL Number MFCD00044261
CAS 17363-02-9
Packaging Glass bottle
Total Trace Metal Impurities 0.01% max.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Name Note 99.99%
Linear Formula (NH4)2PtBr6

For ultra trace analysis

Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.