missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexabromoplatinate(IV), 99.9% (metals basis), Pt 28%

Manufacturer:  Alfa Aesar™ 1261903

 View more versions of this product

Catalog No. AA1261903



Specifications

Specifications

Ammonium hexabromoplatinate(IV)
(metals basis)
MFCD00044261
YLDMEZNZYLWNNL-UHFFFAOYSA-J
diazanium;hexabromoplatinum(2-)
16211461
Powder
Pt 28%
145°C (decomposition)
17363-02-9
Br6H8N2Pt
diazanium;hexabromoplatinum 2-, diammonium hexabromoplatinumdiuide, platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
[NH4+].[NH4+].Br[Pt-2](Br)(Br)(Br)(Br)Br
710.586
710.62
99.9%
Hygroscopic
1g

Chemical Identifiers

17363-02-9
710.586
YLDMEZNZYLWNNL-UHFFFAOYSA-J
16211461
[NH4+].[NH4+].Br[Pt-2](Br)(Br)(Br)(Br)Br
Br6H8N2Pt
MFCD00044261
diazanium;hexabromoplatinum 2-, diammonium hexabromoplatinumdiuide, platinate 2-,hexabromo-,ammonium 1:2 , oc-6-11
diazanium;hexabromoplatinum(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents