Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexabromoplatinate(IV), 99.9% (metals basis), Pt 28%

Chemical Identifiers

Chemical Name or Material Ammonium hexabromoplatinate(IV)
CAS 17363-02-9
Formula Weight 710.62
MDL Number MFCD00044261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1261903

  SDS
alfa aesar™
1261903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1261906

  SDS
alfa aesar™
1261906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 710.62
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 17363-02-9
TSCA TSCA
Physical Form Powder
Melting Point 145°C (dec.)
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Ammonium hexabromoplatinate(IV)
Assay 99.9%
Health Hazard 1 Danger
Sensitivity Hygroscopic
MDL Number MFCD00044261
Quantity 1g
Assay Percent Notes (metals basis)
Concentration Pt 28%
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.