missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium hexabromoosmate(IV), 99.9% (metals basis), Os 26.5 % min

Manufacturer:  Alfa Aesar™ 1264806

 View more versions of this product

Catalog No. AA1264806



Specifications

Specifications

Ammonium hexabromoosmate(IV)
(metals basis)
MFCD00049389
ZPHFWWAVHILXPL-UHFFFAOYSA-J
diazanium;hexabromoosmium(2-)
11136267
Powder
Os 26.5 % min.
Hygroscopic
5g
24598-62-7
Br6H8N2Os
diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
[NH4+].[NH4+].Br[Os-2](Br)(Br)(Br)(Br)Br
705.732
705.73
99.9%
Odorless
Black

Chemical Identifiers

24598-62-7
705.732
ZPHFWWAVHILXPL-UHFFFAOYSA-J
11136267
[NH4+].[NH4+].Br[Os-2](Br)(Br)(Br)(Br)Br
Br6H8N2Os
MFCD00049389
diammonium hexabromoosmiumdiuide, osmate 2-,hexabromo-,ammonium 1:2 , oc-6-11
diazanium;hexabromoosmium(2-)
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents