missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate, Crystal, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A119045KGBL

 View more versions of this product

Catalog No. 18608656

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

540-69-2
5.5 to 7.5
CH5NO2
N.OC=O
63.06
Reagent
0.0001
1
45 kg
VZTDIZULWFCMLS-UHFFFAOYSA-N
formic acid amine
97%
Fiber Drum

Chemical Identifiers

540-69-2
63.06
formic acid amine
CH5NO2
VZTDIZULWFCMLS-UHFFFAOYSA-N
N.OC=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.