Learn More
Ammonium formate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01451730
Description
Ammonium formate is widely used in various organic reactions like Leuckart reaction which involves the reductive amination of aldehydes and ketones. It serves as a buffer in high performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC/MS). It finds application in palladium on carbon (Pd/C) reduction of functional groups. For example, reduction of alkenes to alkanes and formaldehyde to methanol. It is also used to prepare formic acid insitu as well as used to store formic acid by making it as an ammonium salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonium formate | |
540-69-2 | |
Crystalline | |
CH5NO2 | |
MFCD00013103 | |
14,523 | |
Soluble in water,liquid ammonia and alcohol. | |
C(=O)[O-].[NH4+] | |
63.056 | |
CHEBI:63050 | |
98% | |
Hygroscopic |
∼116°C | |
White | |
250 g | |
HCO2NH4 | |
3625095 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
azanium;formate | |
2723923 | |
63.06 | |
Ammonia-like | |
1.28 g/mL |
Chemical Identifiers
540-69-2 | |
63.056 | |
VZTDIZULWFCMLS-UHFFFAOYSA-N | |
2723923 | |
azanium;formate |
CH5NO2 | |
MFCD00013103 | |
ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny | |
CHEBI:63050 | |
C(=O)[O-].[NH4+] |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 208-753-9
RTECSNumber : BQ6650000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only