Please login to your online account to display your discounted pricing

Ammonium formate, 97%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  168615000

 View more versions of this product

Catalog No. AC168615000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0% (total base)
Physical Form Crystals or Crystalline Powder
Formula Weight 63.06
Quantity 500g
Color White
pH 6 to 7 (5% soln. at 20°C)
Fieser 01,38; 13,230; 15,13
CAS 540-69-2
Infrared Spectrum Authentic
Iron (Fe) 20ppm max.
Sulfate 100ppm max.
Heavy Metals (as Pb) 20ppm max.
Packaging Glass bottle
Assay Percent Range 97%
Chlorine (Cl) 50ppm max.
Linear Formula HCO2NH4
Molecular Formula CH5NO2
Chemical Name or Material Ammonium formate
Health Hazard 1 Warning

Specifications

Chemical Name or Material Ammonium formate
Assay ≥96.0% (total base)
Assay Percent Range 97%
Molecular Formula CH5NO2
Linear Formula HCO2NH4
Formula Weight 63.06
Accessories

Warning

SDS & Safety Documents