Please login to your online account to display your discounted pricing

Alfa Aesar™ Ammonium fluoride hydrate, Puratronic™, 99.9975% (metals basis)

Chemical Identifiers

Chemical Name or Material Ammonium fluoride hydrate
CAS 12125-01-8
Purity Grade Puratronic™
Linear Formula NH4F·xH2O (x≈0.5)
Formula Weight 37.04 (Anhydrous)
MDL Number MFCD00011423
UN Number UN2505
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4021614

  SDS
alfa aesar™
4021614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4021622

  SDS
alfa aesar™
4021622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 37.04 (Anhydrous)
Chemical Name or Material Ammonium fluoride hydrate
Odor Odorless
Purity Grade Puratronic™
Physical Form Crystalline Aggregates
Assay Percent Notes (metals basis)
Linear Formula NH4F·xH2O (x≈0.5)
MDL Number MFCD00011423
Color White
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMMONIUM FLUORIDE
RTECS Number BQ6300000
TSCA TSCA
Assay 99.9975%
CAS 12125-01-8
UN Number UN2505
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.