missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium Fluoride, MilliporeSigma™

For analysis EMSURE™ ACS

Manufacturer:  MilliporeSigma 1011640250

 View more versions of this product

Catalog No. M1011640250



Specifications

Specifications

Ammonium fluoride
NH4F
Fluoroammonium
37.04
Ammoniacal
ACS
12125-01-8
UN2505
Soluble in water: 820g/L at 20°C
1.01g/cm3 at 20°C
Plastic Bottle
250g

Chemical Identifiers

12125-01-8
NH4F
Fluoroammonium
Safety and Handling

Safety and Handling

H311: Toxic in contact with skin

331: Toxic if inhaled

H301: Toxic if swallowed

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 235-185-9

Recommended Storage : Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.

Documents
Promotions

Promotions