Please login to your online account to display your discounted pricing

Ammonium Fluoride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$478.60 - $1,051.07

Chemical Identifiers

Chemical Name or Material Ammonium Fluoride
CAS 12125-01-8
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NH4F
Formula Weight 37.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002283

 
avantor performance
069805
Poly Pail 2.5kg Each for $1,051.07

02002284

 
avantor performance
069801
Poly Bottle 500g Each for $478.60
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 37.04
Chemical Name or Material Ammonium Fluoride
Molecular Formula NH4F
CAS 12125-01-8
Packaging Poly Pail
Quantity 2.5kg