Please login to your online account to display your discounted pricing

Ammonium Fluoride, (ACS Reagent Grade), MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0215249590

 View more versions of this product

Catalog No. ICN15249590

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Ammonium Fluoride
CAS 12125-01-8
Purity Grade ACS Reagent
Molecular Formula NH4F
Formula Weight 37.0g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Ammonium Fluoride
CAS 12125-01-8
Purity Grade ACS Reagent
Molecular Formula NH4F
Formula Weight 37.0g/mol
Accessories

Room Temperature