Please login to your online account to display your discounted pricing

Ammonium Fluoride, (ACS Reagent Grade), MP Biomedicals

$82.35 - $167.87

Chemical Identifiers

Chemical Name or Material Ammonium Fluoride
CAS 12125-01-8
Purity Grade ACS Reagent
Molecular Formula NH4F
Formula Weight 37.0g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15249590

 
mp biomedicals
0215249590
500g Each for $167.87

ICN15249580

 
mp biomedicals
0215249580
100g Each for $82.35
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Ammonium Fluoride
CAS 12125-01-8
Purity Grade ACS Reagent
Molecular Formula NH4F
Formula Weight 37.0g/mol
Recommended Storage Room Temperature
Safety and Handling

Room Temperature