missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ammonium fluoride, ACS, 98.0% min, Alfa Aesar

Manufacturer:  Alfa Aesar 11568D9

 View more versions of this product

Catalog No. AA11568D9



Specifications

Specifications

Ammonium fluoride
12125-01-8
NH4F
White crystalline
Crystalline
White
For analysis ACS
98% min.
MFCD00011423
37.04
ACS
5 x 1kg

Chemical Identifiers

12125-01-8
NH4F
MFCD00011423
Safety and Handling

Safety and Handling

H301-H311-H331

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 235-185-9

storageNote1 : Hygroscopic

Documents